missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-dichlorobenzoate, 99%

Product Code. 11450677
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11450677

Brand: Thermo Scientific Alfa Aesar B22489.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2905-69-3 | C8H6Cl2O2 | 205.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-69-3
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD00000606
InChI Key SPJQBGGHUDNAIC-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzoic acid methyl ester, benzoic acid, 2,5-dichloro-, methyl ester, methyl-2,5-dichlorobenzoate, unii-j07555o4he, benzoic acid, dichloro-, methyl ester, pubchem3714, acmc-1bhkt, 2,5-dcbme, rarechem al bf 0427, methyl 2,5-bis chloranyl benzoate
PubChem CID 17947
IUPAC Name methyl 2,5-dichlorobenzoate
SMILES COC(=O)C1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 37°C to 40°C
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 2251582
Formula Weight 205.04
Percent Purity 99%
Chemical Name or Material Methyl 2,5-dichlorobenzoate
compliance-icons compliance-icons
Product Identifier
  • Methyl 2,5-dichlorobenzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 2,5-dichlorobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.