missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-acetamido-5-bromobenzoate, 98%, ACROS Organics™

Product Code. 11965541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11965541 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11965541 Supplier Thermo Scientific Acros Supplier No. 354030050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 138825-96-4
Melting Point 131.0°C to 135.0°C
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H10BrNO3
Linear Formula CH3C(O)NHC6H3(Br)COOCH3
Quantity 5 g
Synonym 2-acetamido-5-bromobenzoic acid methyl ester, methyl 2-acetylamino-5-bromobenzoate, n-acetyl-5-bromoanthranilic acid methyl ester, benzoic acid, 2-acetylamino-5-bromo-, methyl ester, methyl 5-bromo-2-acetamidobenzoate, acmc-20akiv, pubchem3334, methyl2-acetamido-5-bromobenzoate, labotest-bb lt00454800
InChI Key CPARHIBNDSEJGR-UHFFFAOYSA-N
SMILES CC(=O)NC1=C(C=C(C=C1)Br)C(=O)OC
IUPAC Name methyl 2-acetamido-5-bromobenzoate
Molecular Weight (g/mol) 272.098
PubChem CID 736223
Formula Weight 272.1
Percent Purity 98%
Chemical Name or Material Methyl 2-acetamido-5-bromobenzoate
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.