missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(acetylamino)-3-nitrobenzoate, 97%, Thermo Scientific™

Product Code. p-2704380
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10581482

Brand: Thermo Scientific Maybridge RH00481DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 95067-27-9
Molecular Formula C10H10N2O5
Molecular Weight (g/mol) 238.20
MDL Number MFCD00157613
InChI Key PHFRWNCHBPVTJN-UHFFFAOYSA-N
Synonym methyl 2-acetylamino-3-nitrobenzoate, methyl 2-acetamido-3-nitro-benzoate, 2-acetylamino-3-nitro-benzoic acid methyl ester, 2-acetamido-3-nitrobenzoic acid methyl ester, methyl-2-acetylamino-3-nitrobenzoate, methyl2-acetylamino-3-nitrobenzoate, 2-acetylamino-3-nitrobenzoic acid methyl ester
PubChem CID 2782278
IUPAC Name methyl 2-acetamido-3-nitrobenzoate
SMILES COC(=O)C1=C(NC(C)=O)C(=CC=C1)[N+]([O-])=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 238.2
Percent Purity 97%
Chemical Name or Material Methyl 2-(acetylamino)-3-nitrobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.