missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-3-carboxybenzoate, 97%, Thermo Scientific™

Product Code. 10718623
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10718623 250 mg Amber Glass Bottle 250mg
10564403 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10718623 Supplier Thermo Scientific Maybridge Supplier No. MO07276CB
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 253120-47-7
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD06200897
InChI Key UXQCFHRNRAGKOY-UHFFFAOYSA-N
PubChem CID 2794812
IUPAC Name 2-amino-3-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=CC(C(O)=O)=C1N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-amino-3-methoxycarbonyl benzoic acid, methyl 2-amino-3-carboxybenzoate, 2-amino-isophthalic acid monomethyl ester, 3-methoxycarbonyl-2-aminobenzoic acid, 2-amino-isophthalicacidmonomethylester, 2-amino-3-methoxycarbonyl-benzoic acid, pubchem22030, 2-aminoisophthalicacidmonomethylester, 6-carboxy-2-methoxycarbonyl aniline, methyl2-amino-3-carboxybenzoate
Formula Weight 195.17
Percent Purity 97%
Chemical Name or Material Methyl 2-amino-3-carboxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.