missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-3-carboxybenzoate, 97%, Thermo Scientific™

Product Code. p-3111009
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10718623

Brand: Thermo Scientific Maybridge MO07276CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 253120-47-7
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD06200897
InChI Key UXQCFHRNRAGKOY-UHFFFAOYSA-N
PubChem CID 2794812
IUPAC Name 2-amino-3-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=CC(C(O)=O)=C1N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-amino-3-methoxycarbonyl benzoic acid, methyl 2-amino-3-carboxybenzoate, 2-amino-isophthalic acid monomethyl ester, 3-methoxycarbonyl-2-aminobenzoic acid, 2-amino-isophthalicacidmonomethylester, 2-amino-3-methoxycarbonyl-benzoic acid, pubchem22030, 2-aminoisophthalicacidmonomethylester, 6-carboxy-2-methoxycarbonyl aniline, methyl2-amino-3-carboxybenzoate
Formula Weight 195.17
Percent Purity 97%
Chemical Name or Material Methyl 2-amino-3-carboxybenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.