missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-3-methylbenzoate, 98%

Product Code. p-7053778
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15417998

Brand: Thermo Scientific Alfa Aesar H61448.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22223-49-0 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22223-49-0
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD06200918
InChI Key VSFYTPXXMLJNAU-UHFFFAOYSA-N
Synonym methyl 3-methylanthranilate, 2-amino-3-methylbenzoic acid methyl ester, benzoic acid, 2-amino-3-methyl-, methyl ester, methyl 2-amino-3-methylbenzenecarboxylate, 2-amino-3-methyl-benzoic acid methyl ester, methyl2-amino-3-methylbenzoate, pubchem10929, acmc-209ftn, methyl 2-amino-m-toluate, ksc495o6t
PubChem CID 2763406
IUPAC Name methyl 2-amino-3-methylbenzoate
SMILES CC1=CC=CC(=C1N)C(=O)OC

Specifications

Melting Point 25°C to 29°C
Color Pink
Boiling Point 153°C (23 mm)
Quantity 25 g
Formula Weight 165.19
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-amino-3-methylbenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.