missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-4,5-difluorobenzoate, 98%

Product Code. p-7055735
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15491569

Brand: Thermo Scientific Alfa Aesar H64385.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207346-42-7 | C8H7F2NO2 | 187.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207346-42-7
Molecular Formula C8H7F2NO2
Molecular Weight (g/mol) 187.15
MDL Number MFCD09264511
InChI Key VIAQNTRKUPBQKR-UHFFFAOYSA-N
Synonym 2-amino-4,5-difluorobenzoic acid methyl ester, benzoic acid, 2-amino-4,5-difluoro-, methyl ester, pubchem4676, acmc-209fdn, ksc496g7j, methyl 4,5-difluoroanthranilate, methyl2-amino-4,5-difluorobenzoate, methyl 2-amino-4,5-difluoro-benzoate, 2-amino-4,5-difluoro-benzoic acid methyl ester
PubChem CID 18472001
IUPAC Name methyl 2-amino-4,5-difluorobenzoate
SMILES COC(=O)C1=CC(F)=C(F)C=C1N

Specifications

Color White
Quantity 5 g
Beilstein 2834936
Formula Weight 187.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-amino-4,5-difluorobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.