missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-4,5-dimethoxybenzoate, 98%

Product Code. 11420087
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420087 10 g 10g
11430087 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420087 Supplier Thermo Scientific Alfa Aesar Supplier No. B21002.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 26759-46-6 | C10H13NO4 | 211.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26759-46-6
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.22
MDL Number MFCD00014904
InChI Key QQFHCCQSCQBKBG-UHFFFAOYSA-N
Synonym methyl 6-aminoveratrate, methyl 4,5-dimethoxyanthranilate, methyl 2-amino-4,5-dimethoxy benzoate, 2-amino-4,5-dimethoxybenzoic acid methyl ester, 6-aminoveratric acid methyl ester, methyl2-amino-4,5-dimethoxybenzoate, 4,5-dimethoxyanthranilic acid methyl, benzoic acid, 2-amino-4,5-dimethoxy-, methyl ester, methyl-2-amino-4,5-dimethoxybenzoate, 2-amino-4,5-dimethoxy-benzoic acid methyl ester
PubChem CID 611144
IUPAC Name methyl 2-amino-4,5-dimethoxybenzoate
SMILES COC(=O)C1=CC(OC)=C(OC)C=C1N

Specifications

Melting Point 128°C to 130°C
Quantity 10 g
Beilstein 2724754
Formula Weight 211.22
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-4,5-dimethoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.