missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-4,5-dimethoxybenzoate, 98%

Product Code. 11430087
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430087 50 g 50g
11420087 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430087 Supplier Thermo Scientific Alfa Aesar Supplier No. B21002.18
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26759-46-6 | C10H13NO4 | 211.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 26759-46-6
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.22
MDL Number MFCD00014904
InChI Key QQFHCCQSCQBKBG-UHFFFAOYSA-N
Synonym methyl 6-aminoveratrate, methyl 4,5-dimethoxyanthranilate, methyl 2-amino-4,5-dimethoxy benzoate, 2-amino-4,5-dimethoxybenzoic acid methyl ester, 6-aminoveratric acid methyl ester, methyl2-amino-4,5-dimethoxybenzoate, 4,5-dimethoxyanthranilic acid methyl, benzoic acid, 2-amino-4,5-dimethoxy-, methyl ester, methyl-2-amino-4,5-dimethoxybenzoate, 2-amino-4,5-dimethoxy-benzoic acid methyl ester
PubChem CID 611144
IUPAC Name methyl 2-amino-4,5-dimethoxybenzoate
SMILES COC(=O)C1=CC(OC)=C(OC)C=C1N

Spécification

Melting Point 128°C to 130°C
Quantity 50 g
Beilstein 2724754
Formula Weight 211.22
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-4,5-dimethoxybenzoate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.