missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-fluorobenzoate, 98%

Product Code. 11359735
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359735 25 g 25g
11349735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359735 Supplier Thermo Scientific Alfa Aesar Supplier No. L19778.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 319-24-4 | C8H8FNO2 | 169.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 319-24-4
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD03791308
InChI Key PUDDYSBKCDKATP-UHFFFAOYSA-N
Synonym 2-amino-5-fluorobenzoic acid methyl ester, methyl2-amino-5-fluorobenzoate, methyl 5-fluoroanthranilate, methyl 2-amino-5-fluoro-benzoate, benzoic acid, 2-amino-5-fluoro-, methyl ester, 2-amino-5-fluoro benzoic acid methyl ester, 2-amino-5-fluoro-benzoic acid methyl ester, pubchem3558, acmc-209hpn, ksc495g7j
PubChem CID 2783401
IUPAC Name methyl 2-amino-5-fluorobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)F)N

Especificaciones

Melting Point 21°C to 25°C
Density 1.26
Boiling Point 100°C to 102°C (0.5 mmHg)
Flash Point 132°C (269°F)
Quantity 25 g
Beilstein 2803210
Formula Weight 169.16
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-5-fluorobenzoate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.