missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-methoxybenzoate, 96%

Product Code. 15490908
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490908 1 g 1g
15410918 250 mg 250mg
15400918 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490908 Supplier Thermo Scientific Alfa Aesar Supplier No. H54704.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2475-80-1 | C9H11NO3 | 181.19 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly Soluble (1.1 g/L) (25°C) in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2475-80-1
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD09038482
InChI Key MOVBJUGHBJJKOW-UHFFFAOYSA-N
PubChem CID 13052275
IUPAC Name methyl 2-amino-5-methoxybenzoate
SMILES COC(=O)C1=C(N)C=CC(OC)=C1

Specifications

Melting Point 32°C to 33°C
Quantity 1 g
Synonym benzoic acid, 2-amino-5-methoxy-, methyl ester, methyl2-amino-5-methoxybenzoate, 2-amino-5-methoxybenzoic acid methyl ester, methyl 5-methoxyanthranilate, benzoic acid,2-amino-5-methoxy-, methyl ester, methyl 6-amino-m-anisate, acmc-1cnv1, intermediates-zcf02665, 2-amino-5-methoxybenzoic acid,methyl ester, 2-amino-5-methoxy-benzoic acid methyl ester
Solubility Information Slightly Soluble (1.1g/L) (25°C) in water.
Formula Weight 181.19
Percent Purity 96%
Chemical Name or Material Methyl 2-amino-5-methoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.