missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-5-chlorobenzoate, 98%

Product Code. 11476986
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476986 25 g 25g
11486986 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476986 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' B25309.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27007-53-0 | C8H6BrClO2 | 249.488 g/mol

Methyl 2-bromo-5-chlorobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromo-5-chlorobenzoate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27007-53-0
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.488
MDL Number MFCD00144763
InChI Key BIECSXCXIXHDBC-UHFFFAOYSA-N
Synonym 2-bromo-5-chlorobenzoic acid methyl ester, benzoic acid, 2-bromo-5-chloro-, methyl ester, pubchem11238, acmc-1cr8r, ksc494q3f, methyl-2-bromo-5-chlorobenzoate, methyl 2-bromo-5-chloro-benzoate, methyl 2-bromanyl-5-chloranyl-benzoate, 2-bomo-5-chlorobenzoic acid methyl ester
PubChem CID 280500
IUPAC Name methyl 2-bromo-5-chlorobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Cl)Br

Specifications

Melting Point 37°C to 40°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 249.49
Percent Purity 98%
Chemical Name or Material Methyl 2-bromo-5-chlorobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.