missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-5-methoxybenzoate, 98%

Product Code. 11438216
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438216 5 g 5g
11448216 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438216 Supplier Thermo Scientific Alfa Aesar Supplier No. A15322.06
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 35450-36-3 | C9H9BrO3 | 245.07 g/mol

Methyl 2-bromo-5-methoxybenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromo-5-methoxybenzoate is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Avoid contact with strong oxidizing agents. Store in a cool and dark place. Protect from direct light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35450-36-3
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00051594
InChI Key VRTQLDFCPNVQNT-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzoic acid methyl ester, 2-bromo-5-methoxy-benzoic acid methyl ester, methyl-2-bromo-5-methoxybenzoate, benzoic acid, 2-bromo-5-methoxy-, methyl ester, pubchem3965, acmc-1ahhz, methyl 6-bromo-m-anisate, rarechem al bf 1111, attercop-chm at108617, methyl 2-bromo-5-methoxylbenzoate
PubChem CID 2776849
IUPAC Name methyl 2-bromo-5-methoxybenzoate
SMILES COC(=O)C1=CC(OC)=CC=C1Br

Specifications

Density 1.52
Boiling Point 271°C
Flash Point >110°C (230°F)
Refractive Index 1.564
Quantity 5 g
Beilstein 2641378
Solubility Information Not miscible in water.
Formula Weight 245.08
Percent Purity 98%
Chemical Name or Material Methyl 2-bromo-5-methoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.