missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-5-methoxybenzoate, 98%

Product Code. p-7022503
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11438216

Brand: Thermo Scientific Alfa Aesar A15322.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35450-36-3 | C9H9BrO3 | 245.07 g/mol

Methyl 2-bromo-5-methoxybenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromo-5-methoxybenzoate is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Avoid contact with strong oxidizing agents. Store in a cool and dark place. Protect from direct light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35450-36-3
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00051594
InChI Key VRTQLDFCPNVQNT-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzoic acid methyl ester, 2-bromo-5-methoxy-benzoic acid methyl ester, methyl-2-bromo-5-methoxybenzoate, benzoic acid, 2-bromo-5-methoxy-, methyl ester, pubchem3965, acmc-1ahhz, methyl 6-bromo-m-anisate, rarechem al bf 1111, attercop-chm at108617, methyl 2-bromo-5-methoxylbenzoate
PubChem CID 2776849
IUPAC Name methyl 2-bromo-5-methoxybenzoate
SMILES COC(=O)C1=CC(OC)=CC=C1Br

Specifications

Density 1.52
Boiling Point 271°C
Flash Point >110°C (230°F)
Refractive Index 1.564
Quantity 5 g
Beilstein 2641378
Solubility Information Not miscible in water.
Formula Weight 245.08
Percent Purity 98%
Chemical Name or Material Methyl 2-bromo-5-methoxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.