missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromobenzoate, 99%

Product Code. 11489643
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489643 25 g 25g
11499643 100 g 100g
11409653 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489643 Supplier Thermo Scientific Alfa Aesar Supplier No. A12435.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 610-94-6 | C8H7BrO2 | 215.05 g/mol

Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-94-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00016328
InChI Key SWGQITQOBPXVRC-UHFFFAOYSA-N
Synonym methyl o-bromobenzoate, 2-bromobenzoic acid methyl ester, benzoic acid, 2-bromo-, methyl ester, methyl-2-bromobenzoate, methyl bromobenzoate, o-bromobenzoic acid, methyl ester, benzoic acid, o-bromo-, methyl ester, pubchem3956, 2-bromo methyl benzoate, acmc-1atow
PubChem CID 11894
IUPAC Name methyl 2-bromobenzoate
SMILES COC(=O)C1=CC=CC=C1Br

Specifications

Density 1.532
Boiling Point 252°C
Flash Point >110°C (230°F)
Refractive Index 1.559
Quantity 25 g
Beilstein 1862501
Solubility Information Insoluble in water.
Formula Weight 215.05
Percent Purity 99%
Chemical Name or Material Methyl 2-bromobenzoate
compliance-icons
Product Identifier
  • Methyl 2-bromobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Methyl 2-bromobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.