missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chloro-4,4-dimethyl-3-oxopentanoate, 90%, Thermo Scientific™

Product Code. 10405902
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10405902 10 g Amber Glass Bottle 10g
10495892 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10405902 Supplier Thermo Scientific Maybridge Supplier No. KM09505EA
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-33-1
Molecular Formula C8H13ClO3
Molecular Weight (g/mol) 192.639
MDL Number MFCD01570535
InChI Key NGRPVOKPBYTXLT-UHFFFAOYSA-N
PubChem CID 2779362
IUPAC Name methyl 2-chloro-4,4-dimethyl-3-oxopentanoate
SMILES CC(C)(C)C(=O)C(C(=O)OC)Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Linear Formula (CH3)3CC(O)CHClCO2CH3
Quantity 10 g
Synonym 2-chloro-4,4-dimethyl-3-oxo-pentanoic acid methyl ester, methyl 2-chloro-4,4-dimethyl-3-oxo-pentanoate, pentanoic acid,2-chloro-4,4-dimethyl-3-oxo-, methyl ester, pentanoic acid, 2-chloro-4,4-dimethyl-3-oxo-, methyl ester
Formula Weight 192.64
Percent Purity 90%
Chemical Name or Material Methyl 2-chloro-4, 4-dimethyl-3-oxopentanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.