missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluoro-4-hydroxybenzoate, 98%

Product Code. 15468238
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468238 5 g 5g
15478238 250 mg 250mg
15458238 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468238 Supplier Thermo Scientific Alfa Aesar Supplier No. H61675.06
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 197507-22-5 | C8H7FO3 | 170.139 g/mol

Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197507-22-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD09038282
InChI Key WYSPMXSNCAFCFV-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzoic acid methyl ester, 2-fluoro-4-hydroxybenzoic acid methyl ester, benzoic acid, 2-fluoro-4-hydroxy-, methyl ester, methyl-2-fluoro4-hydroxybenzoate, methyl2-fluoro-4-hydroxybenzoate, methyl-2-fluoro-4-hydroxybenzoate, methyl 2-fluoro-4-hydroxy-benzoate, 2-fluoro-4-hyroxybenzoic acid methyl ester
PubChem CID 22463192
IUPAC Name methyl 2-fluoro-4-hydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)O)F

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material Methyl 2-fluoro-4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.