missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxy-4-methoxybenzoate, 98%

Product Code. 11376676
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376676 50 g 50g
11366676 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376676 Supplier Thermo Scientific Alfa Aesar Supplier No. L03855.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5446-02-6 | C9H10O4 | 182.18 g/mol

Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5446-02-6
MDL Number MFCD00008424

Specifications

Melting Point 47°C to 51°C
Boiling Point 134°C (7mmHg)
Molecular Formula C5H10ClNO3
Beilstein 2691644
Synonym 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
Solubility Information Soluble in acetone. Insoluble in water.
InChI Key ZLHFONARZHCSET-UHFFFAOYSA-N
SMILES C(CC(=O)O)C(=O)CN.Cl
IUPAC Name 5-amino-4-oxopentanoic acid;hydrochloride
Molecular Weight (g/mol) 167.59
Quantity 50 g
PubChem CID 123608
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material Methyl 2-hydroxy-4-methoxybenzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.