accessibility menu, dialog, popup

UserName

CH3OOOCH3HO

CAS RN 5446-02-6

IUPAC Name: methyl 2-hydroxy-4-methoxybenzoate
Synonyms: methyl 2-hydroxy-4-methoxybenzoate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: ZICRWXFGZCVTBZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)C=C1O
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results