missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-iodobenzoate, 99+%

Product Code. 10194520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10194520 25 mL Glass bottle 25mL
10085993 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10194520 Supplier Thermo Scientific Acros Supplier No. 249830250
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 610-97-9 | C8H7IO2 | 262.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-97-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.04
MDL Number MFCD00016351
InChI Key BXXLTVBTDZXPTN-UHFFFAOYSA-N
Synonym 2-iodobenzoic acid methyl ester, methyl o-iodobenzoate, methyl-2-iodobenzoate, benzoic acid, 2-iodo-, methyl ester, 2-iodo-benzoic acid methyl ester, methyl2-iodobenzoate, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, pubchem10568, acmc-209mp0
PubChem CID 69132
IUPAC Name methyl 2-iodobenzoate
SMILES COC(=O)C1=CC=CC=C1I

Specifications

Color White
Density 1.7300g/mL
Boiling Point 272.0°C to 274.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4COOCH3
Refractive Index 1.6020 to 1.6040
Quantity 25 mL
Specific Gravity 1.73
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in polar solvents,insoluble in non-polar solvents
Formula Weight 262.04
Percent Purity 99+%
Physical Form Crystalline or Flaky Powder
Chemical Name or Material Methyl 2-iodobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-iodobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.