missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methoxy-5-nitrobenzoate, 98%

Product Code. 11454917
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454917 5 g 5g
11464917 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454917 Supplier Thermo Scientific Alfa Aesar Supplier No. B21863.06
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 34841-11-7 | C9H9NO5 | 211.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34841-11-7
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.173
MDL Number MFCD00673023
InChI Key DOBFJVVTBNTGCW-UHFFFAOYSA-N
Synonym 2-methoxy-5-nitrobenzoic acid methyl ester, methyl2-methoxy-5-nitrobenzoate, methyl-2-methoxy-5-nitrobenzoate, 5-nitro-o-anisic acid methyl ester, pubchem14052, acmc-1adrt, methyl 5-nitro-o-anisate, asischem d13383, rarechem al bf 0877, timtec-bb sbb014757
PubChem CID 7020925
IUPAC Name methyl 2-methoxy-5-nitrobenzoate
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 99°C to 102°C
Quantity 5 g
Formula Weight 211.18
Percent Purity 98%
Chemical Name or Material Methyl 2-methoxy-5-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.