missing translation for 'onlineSavingsMsg'
Learn More

methyl 2-(morpholinosulfonyl)benzoate, 95%, Thermo Scientific™

Product Code. 10759215
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10759215 5 g Amber Glass Bottle 5g
10172744 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10759215 Supplier Thermo Scientific Maybridge Supplier No. CC69622DE
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 502182-56-1
Molecular Formula C12H15NO5S
Molecular Weight (g/mol) 285.31
MDL Number MFCD09879985
InChI Key FJLVVZZMDOHEEM-UHFFFAOYSA-N
Synonym methyl 2-morpholinosulfonyl benzoate, methyl 2-morpholine-4-sulfonyl benzoate, methyl 2-morpholin-4-yl sulphonyl benzoate
PubChem CID 341073
IUPAC Name methyl 2-morpholin-4-ylsulfonylbenzoate
SMILES COC(=O)C1=CC=CC=C1S(=O)(=O)N1CCOCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 285.32
Percent Purity 95%
Chemical Name or Material Methyl 2-(morpholinosulfonyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.