missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-nitro-3,4,5-trimethoxybenzoate, 98%

Product Code. 11392406
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392406 50 g 50g
11382406 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392406 Supplier Thermo Scientific Alfa Aesar Supplier No. L06188.18
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5081-42-5 | C11H13NO7 | 271.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5081-42-5
Molecular Formula C11H13NO7
Molecular Weight (g/mol) 271.225
MDL Number MFCD00007097
InChI Key HCEIEWYPDCDNNU-UHFFFAOYSA-N
Synonym methyl 2-nitro-3,4,5-trimethoxybenzoate, methyl 6-nitro-3,4,5-trimethoxybenzoate, 3,4,5-trimethoxy-2-nitrobenzoic acid methyl ester, benzoic acid, 3,4,5-trimethoxy-2-nitro-, methyl ester, 2-nitro-3,4,5-trimethoxybenzoic acid methyl ester, pubchem22545, opera_id_1985, acmc-1ar8w, hceiewypdcdnnu-uhfffaoysa, methyl 3,5-trimethoxy-2-nitrobenzoate
PubChem CID 78765
IUPAC Name methyl 3,4,5-trimethoxy-2-nitrobenzoate
SMILES COC1=C(C(=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC)OC

Specifications

Melting Point 66°C to 70°C
Quantity 50 g
Beilstein 2702653
Formula Weight 271.23
Percent Purity 98%
Chemical Name or Material Methyl 2-nitro-3,4,5-trimethoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.