Learn More
Methyl 2-pyridyl ketoxime, 97%
Description
Methyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Quantity | 1 g |
| Solubility Information | Slightly soluble in water. |
| IUPAC Name | (2Z)-2-(1-nitrosoethylidene)-1,2-dihydropyridine |
| Formula Weight | 136.15 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl 2-pyridyl ketoxime |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.