missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2E)-[2-(chloromethyl)phenyl)](methoxyimino)acetate, TRC

Product Code. 30011192
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011192 500 mg 500mg
30005622 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011192 Supplier Toronto Research Chemicals Supplier No. TRCM299250500MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11 H12 Cl N O3
Molecular Weight (g/mol) 241.67
IUPAC Name methyl (2E)-2-[2-(chloromethyl)phenyl]-2-methoxyimino-acetate
SMILES CO\N=C(\C(=O)OC)/c1ccccc1CCl

Specifications

Analyte or Component Names Methyl (2E)-[2-(chloromethyl)phenyl](methoxyimino)acetate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C11H12ClNO3/c1-15-11(14)10(13-16-2)9-6-4-3-5-8(9)7-12/h3-6H,7H2,1-2H3/b13-10+
Industry Type Forensics
Quantity 500 mg
Formula Weight 241.0506
Chemical Name or Material Methyl (2E)-[2-(chloromethyl)phenyl](methoxyimino)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.