missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-hydroxyphenyl)propionate, 97%

Product Code. 11464893
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464893 5 g 5g
11474893 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464893 Supplier Thermo Scientific Alfa Aesar Supplier No. A12219.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20349-89-7 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20349-89-7
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00067757
InChI Key YHXYRISRGHSPNV-UHFFFAOYSA-N
Synonym methyl 3-2-hydroxyphenyl propanoate, methyl 3-2-hydroxyphenyl propionate, methyl melilotate, acmc-20aki4, methyl hydroxy-benzenepropionate, methyl 2'-hydroxyhydrocinnamate, methyl 3-hydroxyphenyl propionate, methyl 3 2-hydroxyphenyl propanoate, 2-hydroxybenzenepropanoic acid methyl ester, 3-2-hydroxyphenyl propionic acid methyl ester
PubChem CID 2794569
IUPAC Name methyl 3-(2-hydroxyphenyl)propanoate
SMILES COC(=O)CCC1=CC=CC=C1O

Specifications

Melting Point 40°C to 44°C
Quantity 5 g
Formula Weight 180.21
Percent Purity 97%
Chemical Name or Material Methyl 3- (2-hydroxyphenyl)propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.