missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals

Product Code. 11440491
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440491 5 g 5g
11430491 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440491 Supplier Thermo Scientific Alfa Aesar Supplier No. B20027.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-68-2
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD00018165
InChI Key WICGATIORFSOJL-UHFFFAOYSA-N
PubChem CID 76193
IUPAC Name methyl 3,4-dichlorobenzoate
SMILES COC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 44°C to 46°C
Quantity 5 g
Synonym methyl 3,4-dichlorobenzenecarboxylate, 3,4-dichlorobenzoic acid methyl ester, benzoic acid, 3,4-dichloro-, methyl ester, benzoic acid,3,4-dichloro-, methyl ester, benzoic acid,4-dichloro-, methyl ester, acmc-1ch33, methyldichlorobenzenecarboxylate, methyl 3,4-bis chloranyl benzoate
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material Methyl 3,4-dichlorobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.