missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-dimethoxybenzoate, 98+%, Thermo Scientific™

Product Code. 10133920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
50g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10133920 50g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10133920 Supplier Thermo Scientific Acros Supplier No. 189230500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2150-38-1
Melting Point 58°C to 62°C
Color Brown
Boiling Point 283°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Plastic bottle
Molecular Formula C10H12O4
Linear Formula (CH3O)2C6H3CO2CH3
MDL Number MFCD00008430
Quantity 50 g
Beilstein 10, 396
Synonym methyl veratrate, benzoic acid, 3,4-dimethoxy-, methyl ester, 3,4-dimethoxybenzoic acid methyl ester, veratric acid, methyl ester, methylveratrate, pubchem13009, acmc-1cfay, veratric acid methyl ester, methyl 3,4 dimethoxybenzoate, methyl 3, 4-dimethoxybenzoate
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol,acetone
InChI Key BIGQPYZPEWAPBG-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=C(OC)C(OC)=C1
IUPAC Name methyl 3,4-dimethoxybenzoate
Molecular Weight (g/mol) 196.20
PubChem CID 16522
ChEBI CHEBI:86906
Formula Weight 196.2
Percent Purity 98+%
Physical Form Granular Powder
Chemical Name or Material Methyl 3, 4-dimethoxybenzoate, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.