missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-di-tert-butyl-4-hydroxybenzoate, 98%

Product Code. 11412607
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412607 25 g 25g
11422607 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412607 Supplier Thermo Scientific Alfa Aesar Supplier No. A14837.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2511-22-0 | C16H24O3 | 264.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2511-22-0
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.37
MDL Number MFCD00017253
InChI Key UPVYFJALDJUSOV-UHFFFAOYSA-N
Synonym methyl 3,5-di-tert-butyl-4-hydroxybenzoate, benzoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy-, methyl ester, 3,5-di-tert-butyl-4-hydroxybenzoic acid methyl ester, 3,5-bis 1,1-dimethylethyl-4-hydroxy-benzoic acid ethyl ester, acmc-20akhp, methyl 3,5-bis-tert-butyl-4-hydroxybenzoate, benzoic acid, 3,5-di-tert-butyl-4-hydroxy-, methyl ester, methyl 3,5-di-tert.butyl-4-hydroxylbenzoate, methyl 3,5-bis tert-butyl-4-hydroxybenzoate
PubChem CID 75639
IUPAC Name methyl 3,5-ditert-butyl-4-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 164°C to 166°C
Quantity 25 g
Beilstein 2218125
Formula Weight 264.37
Percent Purity 98%
Chemical Name or Material Methyl 3,5-di-tert-butyl-4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.