missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dichlorobenzoate, 98%, Thermo Scientific™

Product Code. 11441000
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441000 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441000 Supplier Thermo Scientific Alfa Aesar Supplier No. B25311.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-67-1
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD00032710
InChI Key BTEVDFJXGLQUDS-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzoic acid methyl ester, benzoic acid, 3,5-dichloro-, methyl ester, unii-4gim06ee3p, 4gim06ee3p, methyl-3,5-dichlorobenzoate, maybridge1_004559, acmc-1cd9x, dsstox_cid_22146, dsstox_rid_79937, dsstox_gsid_42146
PubChem CID 76192
IUPAC Name methyl 3,5-dichlorobenzoate
SMILES COC(=O)C1=CC(Cl)=CC(Cl)=C1

Specifications

Melting Point 58°C to 60°C
Quantity 25 g
Formula Weight 205.04
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dichlorobenzoate
compliance-icons
Product Identifier
  • Methyl 3,5-dichlorobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Methyl 3,5-dichlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.