missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dichlorobenzoate, Tech., Thermo Scientific™

Product Code. 10557084
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10557084 25 g 25g
10025002 1 g 1g
10310723 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10557084 Supplier Thermo Scientific Maybridge Supplier No. KM02512EB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2905-67-1
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD00032710
InChI Key BTEVDFJXGLQUDS-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzoic acid methyl ester, benzoic acid, 3,5-dichloro-, methyl ester, unii-4gim06ee3p, 4gim06ee3p, methyl-3,5-dichlorobenzoate, maybridge1_004559, acmc-1cd9x, dsstox_cid_22146, dsstox_rid_79937, dsstox_gsid_42146
PubChem CID 76192
IUPAC Name methyl 3,5-dichlorobenzoate
SMILES COC(=O)C1=CC(Cl)=CC(Cl)=C1

Specifications

Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 205.04
Percent Purity Tech.
Grade Technical
Chemical Name or Material Methyl 3, 5-dichlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.