missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-Dichlorobenzoate, TRC

Product Code. 30134570
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134570 1 g 1g
30134458 500 mg 500mg
30003372 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134570 Supplier Toronto Research Chemicals Supplier No. TRCM3263431G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-67-1
Molecular Formula C8 H6 Cl2 O2
Molecular Weight (g/mol) 205.04
Synonym Benzoic acid, 3,5-dichloro-, methyl ester, 3,5-Dichlorobenzoic acid methyl ester, Methyl 3,5-dichlorobenzoate
IUPAC Name methyl 3,5-dichlorobenzoate
SMILES COC(=O)c1cc(Cl)cc(Cl)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H6Cl2O2/c1-12-8(11)5-2-6(9)4-7(10)3-5/h2-4H,1H3
Formula Weight 203.9745
Chemical Name or Material 3,5-Dichlorobenzoic acid methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.