missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-Dichlorobenzoate, TRC

Product Code. 30003372
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003372 100 mg 100mg
30134570 1 g 1g
30134458 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003372 Supplier Toronto Research Chemicals Supplier No. TRCM326343100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-67-1
InChI Formula InChI=1S/C8H6Cl2O2/c1-12-8(11)5-2-6(9)4-7(10)3-5/h2-4H,1H3
IUPAC Name methyl 3,5-dichlorobenzoate
Molecular Formula C8 H6 Cl2 O2
Molecular Weight (g/mol) 205.04
Chemical Name or Material 3,5-Dichlorobenzoic acid methyl ester

Specifications

Quantity 100 mg
Synonym Benzoic acid, 3,5-dichloro-, methyl ester, 3,5-Dichlorobenzoic acid methyl ester, Methyl 3,5-dichlorobenzoate
SMILES COC(=O)c1cc(Cl)cc(Cl)c1
Formula Weight 203.9745
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.