missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dimethoxybenzoate, 98%

Product Code. 11450383
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450383 50 g 50g
11440383 10 g 10g
11460383 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450383 Supplier Thermo Scientific Alfa Aesar Supplier No. A11486.18
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2150-37-0 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-37-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008432
InChI Key YXUIOVUOFQKWDM-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzoic acid methyl ester, benzoic acid, 3,5-dimethoxy-, methyl ester, 3,5-dimethoxybenzoic acid methyl, methyl 3,5-bis methyloxy benzoate, acmc-1ccvs, rarechem al bf 0065, methyl 3,5-dimethoxybnezoate, ksc489q8p, ethyl 3,5-dimethoxybenzoate, methyl 3,5-dimethoxybenzoate
PubChem CID 75074
IUPAC Name methyl 3,5-dimethoxybenzoate
SMILES COC(=O)C1=CC(OC)=CC(OC)=C1

Specifications

Melting Point 40°C to 43°C
Boiling Point 298°C
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 2693126
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dimethoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.