missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dimethoxybenzoate, 98%

Product Code. p-7081483
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11440383

Brand: Thermo Scientific Alfa Aesar A11486.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2150-37-0 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-37-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008432
InChI Key YXUIOVUOFQKWDM-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzoic acid methyl ester, benzoic acid, 3,5-dimethoxy-, methyl ester, 3,5-dimethoxybenzoic acid methyl, methyl 3,5-bis methyloxy benzoate, acmc-1ccvs, rarechem al bf 0065, methyl 3,5-dimethoxybnezoate, ksc489q8p, ethyl 3,5-dimethoxybenzoate, methyl 3,5-dimethoxybenzoate
PubChem CID 75074
IUPAC Name methyl 3,5-dimethoxybenzoate
SMILES COC(=O)C1=CC(OC)=CC(OC)=C1

Specifications

Melting Point 40°C to 43°C
Boiling Point 298°C
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 2693126
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dimethoxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.