missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-methylbenzoate, 97%, Thermo Scientific Chemicals

Product Code. 10883011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10883011 5 g Glass bottle 5g
10149699 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10883011 Supplier Thermo Scientific Acros Supplier No. 371660050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 18595-18-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00025206
InChI Key YEPWCJHMSVABPQ-UHFFFAOYSA-N
Synonym 3-amino-4-methylbenzoic acid methyl ester, methyl 3-amino-4-methyl-benzoate, benzoic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-p-toluate, methyl 4-methyl-3-aminobenzoate, 3-amino-p-toluic acid methyl ester, 3-amino-4-methyl-benzoic acid methyl ester, 3-amino-4-methyl benzoic acid methyl ester, pubchem2351, pubchem10930
PubChem CID 337778
IUPAC Name methyl 3-amino-4-methylbenzoate
SMILES COC(=O)C1=CC=C(C)C(N)=C1

Tekniske data

Melting Point 114.0°C to 117.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3(NH2)C6H3COOCH3
Quantity 5 g
Formula Weight 165.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-amino-4-methylbenzoate
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Methyl 3-amino-4-methylbenzoate
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.