missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-4-hydroxybenzoate, 98%

Product Code. 15413897
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413897 50 g 50g
15403897 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413897 Supplier Thermo Scientific Alfa Aesar Supplier No. H30222.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 29415-97-2 | C8H7BrO3 | 231.05 g/mol

A reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29415-97-2
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD06203850
InChI Key RKUNSPWAQIUGEZ-UHFFFAOYSA-N
Synonym methyl3-bromo-4-hydroxybenzoate, 3-bromo-4-hydroxybenzoic acid methyl ester, benzoic acid, 3-bromo-4-hydroxy-, methyl ester, 2-bromo-4-methoxycarbonyl phenol, methyl 4-hydroxy-3-bromobenzoate, methyl-3-bromo-4-hydroxybenzoate, methyl 3-bromo-4-hydroxy-benzoate, methyl 3-bromo-4-hydroxybenzoate, methyl 3-bromanyl-4-oxidanyl-benzoate, 3-bromo-4-hydroxy-benzoic acid methyl ester
PubChem CID 4778958
IUPAC Name methyl 3-bromo-4-hydroxybenzoate
SMILES COC(=O)C1=CC(Br)=C(O)C=C1

Specifications

Melting Point 108°C to 110°C
Quantity 50 g
Formula Weight 231.05
Percent Purity 98%
Chemical Name or Material Methyl 3-bromo-4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.