missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-4-methoxybenzoate, 98%

Product Code. 11392359
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392359 10 g 10g
11382359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11392359 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H27297.09
 Restricted Item
Estimated Shipment: 01-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 35450-37-4 | C9H9BrO3 | 245.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 35450-37-4
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00210463
InChI Key ZREVPFANJBZHEU-UHFFFAOYSA-N
Synonym 3-bromo-4-methoxybenzoic acid methyl ester, methyl3-bromo-4-methoxybenzoate, benzoic acid, 3-bromo-4-methoxy-, methyl ester, acmc-209igx, methyl 3-bromo-p-anisate, rarechem al bf 0247, methyl-3-bromo-4-methoxybenzoate, 3-bromo-p-anisic acid methyl ester, #, methyl 3-bromo-4-methyloxy benzoate
PubChem CID 611662
IUPAC Name methyl 3-bromo-4-methoxybenzoate
SMILES COC(=O)C1=CC(Br)=C(OC)C=C1

Specifica

Melting Point 95°C to 98°C
Quantity 10 g
Formula Weight 245.08
Percent Purity 98%
Chemical Name or Material Methyl 3-bromo-4-methoxybenzoate

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.