missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-4-methylbenzoate, 99%

Product Code. 11435807
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11435807

Brand: Thermo Scientific Alfa Aesar B24372.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104901-43-1 | C9H9BrO2 | 229.073 g/mol

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104901-43-1
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00144769
InChI Key MASRAGFWFYHMFI-UHFFFAOYSA-N
Synonym 3-bromo-4-methylbenzoic acid methyl ester, methyl 3-bromo-p-toluate, methyl3-bromo-4-methylbenzoate, benzoic acid, 3-bromo-4-methyl-, methyl ester, 3-bromo-p-toluic acid methyl ester, 3-bromo-4-methyl methyl benzoate, 3-bromo-4-methyl-benzoic acid methyl ester, pubchem3967, acmc-2098dw, methyl 3-bromo4-methylbenzoate
PubChem CID 7015747
IUPAC Name methyl 3-bromo-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)Br

Specifications

Quantity 5 g
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 229.07
Percent Purity 99%
Chemical Name or Material Methyl 3-bromo-4-methylbenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.