missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-5-hydroxybenzoate, 97%, Thermo Scientific Chemicals

Product Code. 15468177
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468177 1 g 1g
15478177 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468177 Supplier Thermo Scientific Alfa Aesar Supplier No. H51724.03
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 192810-12-1 | Molecular Formula: C8H7BrO3 | Molecular Weight: 231.045 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192810-12-1
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD10566800
InChI Key KRTBWIIGEJIUSA-UHFFFAOYSA-N
Synonym 3-bromo-5-hydroxybenzoic acid methyl ester, acmc-209f1w, methyl3-bromo-5-hydroxybenzoate, methyl 3-bromo-5-hydroxylbenzoate, 3-bromo-5-hydroxy-benzoic acid methyl ester, benzoic acid,3-bromo-5-hydroxy-,methyl ester, benzoic acid, 3-bromo-5-hydroxy-, methyl ester
PubChem CID 53216607
IUPAC Name methyl 3-bromo-5-hydroxybenzoate
SMILES COC(=O)C1=CC(=CC(=C1)Br)O

Specifications

Melting Point 130°C to 135°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 231.05
Percent Purity 97%
Chemical Name or Material Methyl 3-bromo-5-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.