missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chloro-4-methylbenzoate, Thermo Scientific™

Product Code. 10682073
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10682073 1 g 1g
10160892 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10682073 Supplier Thermo Scientific Maybridge Supplier No. TL00703DA
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 56525-63-4
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
InChI Key KTFQDZCNPGFKAH-UHFFFAOYSA-N
Synonym methyl3-chloro-4-methylbenzoate, methyl-3-chloro-4-methylbenzoate, methyl 3-chloro-p-toluate, 2-chloro-4-methoxycarbonyl toluene, 3-chloro-4-methylbenzoic acid methyl ester, benzoic acid, 3-chloro-4-methyl-, methyl ester, pubchem10935, acmc-209lsv, ksc495i9j, methyl-3-chloro-4-metylbenzoate
PubChem CID 2801405
IUPAC Name methyl 3-chloro-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)Cl

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material Methyl 3-chloro-4-methylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.