missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(chlorosulfonyl)-4-methoxybenzoate, 97%, Thermo Scientific™

Product Code. 11336885
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11336885 5 g 5g
10181122 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336885 Supplier Thermo Scientific Maybridge Supplier No. MO00348DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 192323-12-9
Molecular Formula C9H9ClO5S
Molecular Weight (g/mol) 264.68
MDL Number MFCD03407355
InChI Key FKAIQWMEWVOGDP-UHFFFAOYSA-N
Synonym methyl 3-chlorosulfonyl-4-methoxybenzoate, methyl 3-chlorosulfonyl-4methoxybenzoate, 2-chlorosulphonyl-4-methoxycarbonyl anisole, methyl 3-chlorosulfonyl-4-methyloxy benzoate, methyl-3-chlorosulfonyl-4-methoxybenzoate, 2-methoxy-5-methoxycarbonyl benzenesulphonyl chloride, benzoic acid,3-chlorosulfonyl-4-methoxy-, methyl ester
PubChem CID 2794613
SMILES COC(=O)C1=CC(=C(OC)C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CH3OC6H3(C(O)OCH3)S(Cl)O2
Quantity 5 g
IUPAC Name methyl 3-(chlorosulfonyl)-4-methoxybenzoate
Formula Weight 264.68
Percent Purity 97%
Chemical Name or Material Methyl 3-(chlorosulfonyl)-4-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.