missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-fluoro-4-methoxybenzoate, 95%, Thermo Scientific™

Product Code. 11319985
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319985 10 g 10g
11329985 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319985 Supplier Thermo Scientific Alfa Aesar Supplier No. H31994.09
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-30-2
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.17
MDL Number MFCD00215839
InChI Key MIKOKYAREMINFF-UHFFFAOYSA-N
Synonym benzoic acid, 3-fluoro-4-methoxy-, methyl ester, methyl 3-fluoro-4-methyloxy benzoate, 3-fluoro-4-methoxybenzoic acid methyl ester, pubchem3569, acmc-1ck9v, rarechem al bf 0055, #
PubChem CID 592848
IUPAC Name methyl 3-fluoro-4-methoxybenzoate
SMILES COC(=O)C1=CC(F)=C(OC)C=C1

Specifications

Melting Point 69°C to 73°C
Quantity 10 g
Formula Weight 184.17
Percent Purity 95%
Chemical Name or Material Methyl 3-fluoro-4-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.