missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxy-4-nitrobenzoate, 98%

Product Code. p-7022510
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11498226

Brand: Thermo Scientific Alfa Aesar A15329.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 713-52-0 | C8H7NO5 | 197.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 713-52-0
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00016997
InChI Key UEGCRFNWTGYVKX-UHFFFAOYSA-N
Synonym benzoic acid, 3-hydroxy-4-nitro-, methyl ester, 3-hydroxy-4-nitrobenzoic acid methyl ester, 3-hydroxy-4-nitro-benzoic acid methyl ester, methyl 3-hydroxy-4-nitro-benzoate, methyl3-hydroxy-4-nitrobenzoate, pubchem20121, acmc-209oia, ksc379e5n, 5-methoxycarbonyl-2-nitrophenol, methyl3-hydroxy-4-nitronemzoate
PubChem CID 294866
IUPAC Name methyl 3-hydroxy-4-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])O

Specifications

Melting Point 89°C to 92°C
Quantity 5 g
Beilstein 2583832
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material Methyl 3-hydroxy-4-nitrobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.