missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-iodo-4-methoxybenzoate, 98%

Product Code. 10422044 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10422044 25 g Glass bottle 25g
10693595 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10422044 Supplier Thermo Scientific Acros Supplier No. 399090250
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35387-93-0 | C9H9IO3 | 292.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35387-93-0
Molecular Formula C9H9IO3
Molecular Weight (g/mol) 292.07
MDL Number MFCD00052925
InChI Key GHNGBFHLUOJHKP-UHFFFAOYSA-N
Synonym 3-iodo-4-methoxy-benzoic acid methyl ester, 3-iodo-4-methoxybenzoic acid methyl ester, benzoic acid, 3-iodo-4-methoxy-, methyl ester, methyl3-iodo-4-methoxybenzoate, methyl 3-iodo-p-anisate, acmc-1agb4, ksc495q3h, methyl-3-iodo-4-methoxybenzoate, methyl 3-iodo-4-methoxy-benzoate, 3-iodo-p-anisic acid methyl ester
PubChem CID 316980
IUPAC Name methyl 3-iodo-4-methoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C(I)=C1

Specifications

Melting Point 93°C to 95°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(I)COOCH3
Quantity 25 g
Formula Weight 292.07
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-iodo-4-methoxybenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-iodo-4-methoxybenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.