missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxy-4-methylbenzoate, 98%

Product Code. 11485337
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485337 25 g 25g
11465337 1 g 1g
11475337 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485337 Supplier Thermo Scientific Alfa Aesar Supplier No. B22695.14
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3556-83-0 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3556-83-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00082710
InChI Key LLEXCSBUSVRBCA-UHFFFAOYSA-N
Synonym 3-methoxy-4-methylbenzoic acid methyl ester, methyl 3-methoxy-p-toluate, 4-methyl-m-anisic acid methyl ester, benzoic acid, 3-methoxy-4-methyl-, methyl ester, pubchem13650, methyl 4-methyl-m-anisate, acmc-1cs22, methyl3-methoxy-4-methylbenzoate, methyl 3-methoxy-4-methyl-benzoate, 3-methoxy-p-toluic acid methyl ester
PubChem CID 591123
IUPAC Name methyl 3-methoxy-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)OC

Specifications

Melting Point 47°C to 50°C
Boiling Point 119°C to 120°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5295
Quantity 25 g
Beilstein 1948339
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material Methyl 3-methoxy-4-methylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.