missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-nitrobenzoate, 98+%

Product Code. 11492138
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492138 250 g 250g
11482138 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492138 Supplier Thermo Scientific Alfa Aesar Supplier No. A15155.30
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 618-95-1 | C8H7NO4 | 181.147 g/mol

Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-95-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007250
InChI Key AXLYJLKKPUICKV-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
PubChem CID 69260
IUPAC Name methyl 3-nitrobenzoate
SMILES COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 77°C to 80°C
Boiling Point 279°C
Quantity 250 g
Beilstein 392449
Solubility Information Insoluble in water.
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material Methyl 3-nitrobenzoate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.