missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals

Product Code. 10076022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10076022 5 g Glass bottle 5g
10798833 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10076022 Supplier Thermo Scientific Acros Supplier No. 381920050
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22621-41-6
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.15
MDL Number MFCD00272263
InChI Key NIBVYEHAFBEVFI-UHFFFAOYSA-N
Synonym methyl 2-hydroxy-3-nitrobenzoate, methyl 3-nitrosalicylate, benzoic acid, 2-hydroxy-3-nitro-, methyl ester, 2-hydroxy-3-nitrobenzoic acid methyl ester, 2-hydroxy-3-nitro-benzoic acid methyl ester, salicylic acid, 3-nitro-, methyl ester, methyl-3-nitrosalicylat, methyl-m-nitrosalicylate, acmc-209fxo, ksc490c4l
PubChem CID 596904
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O

Specifications

Melting Point 127°C to 132°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NO2C6H3(OH)COOCH3
Quantity 5 g
Formula Weight 197.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-nitrosalicylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-nitrosalicylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.