missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Acetamido-5-bromo-2-methoxybenzoate, TRC

Product Code. 30132711
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132711 2.5 g 2.5g
17917334 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132711 Supplier Toronto Research Chemicals Supplier No. TRCM2750452.5G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4093-34-9
Molecular Formula C11H12BrNO4
Molecular Weight (g/mol) 302.12
Synonym 4-Acetamido-5-bromo-o-anisic Acid Methyl Ester, 4-(Acetylamino)-5-bromo-2-methoxy-benzoic Acid Methyl Ester, Bromopride Impurity B
IUPAC Name methyl 4-acetamido-5-bromo-2-methoxybenzoate
SMILES COC(=O)c1cc(Br)c(NC(=O)C)cc1OC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C11H12BrNO4/c1-6(14)13-9-5-10(16-2)7(4-8(9)12)11(15)17-3/h4-5H,1-3H3,(H,13,14)
Formula Weight 300.995
Chemical Name or Material Methyl 4-Acetamido-5-bromo-2-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.