missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-acetamido-5-chloro-2-methoxybenzoate, 98%, Thermo Scientific™

Product Code. 11459886
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459886 10 g 10g
11469886 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459886 Supplier Thermo Scientific Alfa Aesar Supplier No. B20869.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4093-31-6
Molecular Formula C11H12ClNO4
Molecular Weight (g/mol) 257.67
MDL Number MFCD00048191
InChI Key OUEXNQRVYGYGIK-UHFFFAOYSA-N
Synonym methyl 4-acetamido-5-chloro-o-anisate, unii-3d7z5o4uxa, benzoic acid, 4-acetylamino-5-chloro-2-methoxy-, methyl ester, 3d7z5o4uxa, methyl 4-acetylamino-5-chloro-2-methoxybenzoate, methyl 5-chloro-4-acetamido-2-methoxybenzoate, 2-methoxy-4-acetylamino-5-chloro methyl benzoate, 4-acetamido-5-chloro-2-methoxybenzoic acid methyl ester, acmc-1afsv, dsstox_cid_31529
PubChem CID 77721
SMILES COC(=O)C1=CC(Cl)=C(NC(C)=O)C=C1OC

Specifications

Melting Point 153°C to 156°C
Quantity 10 g
IUPAC Name methyl 5-chloro-4-acetamido-2-methoxybenzoate
Formula Weight 257.67
Percent Purity 98%
Chemical Name or Material Methyl 4-acetamido-5-chloro-2-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.